Learn More
Stoelting™ Zero Maze
SDP
Catalog No. 10000309 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Rat
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal
10-000-309 Mouse
10-000-331 Rat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-309 Supplier Stoelting™ Supplier No. 60190
Add to Cart
Add to Cart
Stoelting™ Zero-Maze is a modification of the Elevated Plus-Maze which incorporates both traditional and novel ethological measures in the analysis of anxiety.
Designed with an elevated circular platform with two opposite enclosed quadrants and two open quadrants to eliminate ambiguity in interpretation of time spent on the central square of the traditional design
Specifications
For Use With (Application) Neuroscience
Animal Mouse