Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zero Oxygen Standard for Calibration of Dissolved Oxygen Meters, Reagents

Catalog No. 01333349
Change view
Click to view available options
Quantity:
120 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-349 120 mL
01-333-348 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-333-349 Supplier Reagents Holdings Llc Supplier No. CZ129700120A
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zero Oxygen Standard for Calibration of Dissolved Oxygen Meters, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Zero Oxygen Standard for Calibration of Dissolved Oxygen Meters
Packaging Poly Bottle
CAS 7732-18-5, 7757-83-7
Specific Gravity 1.0311
Physical Form Liquid
Color Colorless
Boiling Point 101°C
Solubility Completely miscible
Quantity 120 mL
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.