Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meccanica GM Divisione Medicale ZERO ZERO Prefilled Formalin Container, 500 mL

Catalog No. 22050002
Change view
Click to view available options
Closure Type:
Screw Thread Cap (Slotted)
Screw Thread Cap (Unslotted)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type
22-050-002 Screw Thread Cap (Slotted)
22-050-001 Screw Thread Cap (Unslotted)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-050-002 Supplier Meccanica GM Divisione Medicale ZERO Supplier No. 50002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meccanica GM Divisione Medicale ZERO Prefilled Formalin Container, 500 mL protects against exposure to harmful chemicals and vapors during routine fixation of specimens in formalin.

  • Reduces or eliminates exposure to formalin and harmful vapors
  • Comes in slotted and unslotted variations
  • Security seal
  • Dual Chamber keeps formalin trapped until container is activated

Specifications

Product Type Pre-filled Specimen Container
Closure Included Yes
Graduated No
Material Polypropylene
Shape Cylindrical
Sterility Non-sterile
Capacity (Metric) 500 mL
Closure Material Plastic
Closure Type Screw Thread Cap (Slotted)
Description Slotted
Diameter (Metric) 9.525 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.