Learn More
Hach Company Hach Zero Solution Less Than 0.05 mg/L TOC, 4 L
Supplier:  Hach Company 5847700
Encompass
Zero Solution less than 0.05 mg/L TOC, 4 L is a certified low-total organic carbon (TOC) solution intended for use as a calibration blank and verification standard for TOC analyzers. It provides a near-zero TOC baseline for instrument calibration, method blanks, and analytical quality control. Consult the product SDS for handling and storage instructions.
  • Provides TOC below 0.05 mg/L.
  • Packaged in a 4 L bottle.
  • Intended for calibration, verification, and method blanks for TOC analysis.
  • Safety Data Sheet (SDS) available; follow handling and storage instructions.
  • Manufacturer lead time/availability indicated on product page.
Catalog No. NC2373299