Learn More
Electron Microscopy Sciences Zero Stat Anti-Static Gun
SDP
Catalog No. NC1350621
Encompass_Preferred
This device operates on a piezoelectric effect to emit positive (trigger squeeze) and negative (trigger release) charges to neutralize static on virtually any surface. The gun remits no radioactivity and it does not require electricity or batteries. The unit measures 4 x 6".
Catalog No. NC1350621 Supplier Electron Microscopy Sciences Supplier No. 60610
This device operates on a piezoelectric effect to emit positive (trigger squeeze) and negative (trigger release) charges to neutralize static on virtually any surface. The gun remits no radioactivity and it does not require electricity or batteries. The unit measures 4 x 6".