Learn More
Cayman Chemical ZeroC™ Dried Plant Matrix, 10 g
SDP
Catalog No. 502918613
Encompass_Preferred
ZeroC™ is a novel cannabinoid-free Cannabis reference matrix designed to meet the rigorous demands of laboratories, researchers, and government agencies.; It serves as an essential tool in the field of Cannabis science for quality control, method validation, and accurate analyte quantification.; ZeroC™ allows laboratories to reliably check the effectiveness of their analytical instruments, validate their test methods, and combat the problem of "laboratory shopping" by establishing a standardized blank matrix.; This Cannabis plant-based reference material serves as a comprehensive solution for investigating common interferences associated with testing cannabinoids, pesticides, mycotoxins, heavy metals, terpenes, residual solvents, and microbials.; ZeroC™ is a naturally produced and non-extracted form of Cannabis, void of any major or minor phytocannabinoids including THC, CBN, CBG, CBD, and THCv.
Catalog No. 50-291-8613 Supplier Cayman Chemical Supplier No. 3933610 G
ZeroC™ is a novel cannabinoid-free Cannabis reference matrix designed to meet the rigorous demands of laboratories, researchers, and government agencies.; It serves as an essential tool in the field of Cannabis science for quality control, method validation, and accurate analyte quantification.; ZeroC™ allows laboratories to reliably check the effectiveness of their analytical instruments, validate their test methods, and combat the problem of "laboratory shopping" by establishing a standardized blank matrix.; This Cannabis plant-based reference material serves as a comprehensive solution for investigating common interferences associated with testing cannabinoids, pesticides, mycotoxins, heavy metals, terpenes, residual solvents, and microbials.; ZeroC™ is a naturally produced and non-extracted form of Cannabis, void of any major or minor phytocannabinoids including THC, CBN, CBG, CBD, and THCv.