Learn More
Andwin Scientific ZEROSTAT ANTI-STATIC
SDP
Catalog No. NC9657211
Encompass_Preferred
The Zerostat anti-static instrument helps in maintaining film, glass, and plasticware free of dust and lint. This product incorporates the latest advancement with a unique Piezo Crystal device that has been proven worldwide in neutralizing static electricity found in a variety of items located in the laboratory. This instrument features portability and versatility. It does not require batteries, electrical power supply, or refills to operate.
Catalog No. NC9657211 Supplier Andwin Scientific Supplier No. Z108812
The Zerostat anti-static instrument helps in maintaining film, glass, and plasticware free of dust and lint. This product incorporates the latest advancement with a unique Piezo Crystal device that has been proven worldwide in neutralizing static electricity found in a variety of items located in the laboratory. This instrument features portability and versatility. It does not require batteries, electrical power supply, or refills to operate.