Learn More
Andwin Scientific Zerostat Antistatic Gun, piezoelectric effect to emit positive (trigger squeeze) and negative (trigger release) charges
SDP
Catalog No. 50727509
Encompass_Preferred
The Zerostat Antistatic Gun works on a piezoelectric effect to emit positive (trigger squeeze) and negative (trigger release) charges to neutralize static on virtually any surface. This product does not need any electricity or batteries to operate.
Catalog No. 50-727-509 Supplier Andwin Scientific Supplier No. 60610
The Zerostat Antistatic Gun works on a piezoelectric effect to emit positive (trigger squeeze) and negative (trigger release) charges to neutralize static on virtually any surface. This product does not need any electricity or batteries to operate.