Learn More
AdipoGen Zerumbone
SDP
Supplier:  AdipoGen AGCN20004M050
Encompass_Preferred
Chemical. CAS 471-05-6. Formula C15H22O. MW 218.3. Isolated from the underground parts of Curcuma zerumbet. Chemopreventive probe. Apoptosis inducer. Potent anti-tumor compound. Anti-inflammatory activity by downregulating COX-2 and iNOS expression via modulation of NF-kappaB activation. Antinociceptive. Probe to study chemokine receptors functioning. Potential therapeutic agent for osteoporosis and cancer-associated bone loss. Anti diabetic nephropathy
Catalog No. 50-256-2701