Learn More
SOLVENTUM 3M™ Zeta Plus™ Encapsulated Filter Capsules with ZB Grade Media 4/pk
Supplier:  SOLVENTUM 7100032799
Encompass_Preferred
3M™ Zeta Plus™ Encapsulated Filter Capsules with ZB Grade Media extend depth filtration capacity and enhances the protection of downstream membranes thereby extending filter life. 3M™ Zeta Plus™ ™ Encapsulated Filter Capsules with ZB Grade Media are part of a family of advanced dual zone depth filters designed to provide optimal clarification of bioprocess, biological and pharmaceutical fluids. 4/pk
Catalog No. NC0088165