Description
- CP Series depth filtration media for clarification of pharmaceutical, biological and bioprocess fluids.
- Combines mechanical depth filtration with charge-assisted adsorption of fine contaminants.
- Provides high contaminant holding capacity for clarification applications.
- Helps reduce particulate load on downstream filtration steps.
- Disc format supports media evaluation and small-scale filtration studies.
Specifications
Specifications
| Grade | 10CP |
| Quantity | 240/Cs. |
| Type | Filter Media Disc |