Learn More
Thermo Scientific Zeta Plus™ EZP Series, 340 cm2
Description
- Two layers of complementary porosities provide high contaminant holding capacity, economical filtration and reliable particle reduction
- FDA Drug Master File and USP <88> Class VI Biological Reactivity Tests eases validation and regulatory submissions
- Self-contained, single-use capsules are available in three convenient sizes for scale-up and benchtop studies
- Low hold-up volume for minimal fluid loss
- Novel shape maintains the same lenticular flow path as larger capsules and cartridges
- High contaminant holding capacity due to two layers of complementary porosities provides economical filtration and reliable particle reduction
Specifications
Specifications
| Disposable | Single-use |
| Material (Membrane) | Cellulose |
| Sterility | Non-sterile |
| Grade | 30SP |
| Quantity | 1 Ea. |
| Type | Filter Capsule |