Learn More
Thermo Scientific™ Zeta Plus™ LA Series Media Sheet, 40.15 x 40.15 in., No Holes
Catalog No. 02557805 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
40/Cs.
Catalog No. Quantity
02-557-805 40/Cs.
1 options
The brand will transition from 3M to Thermo Scientific
Catalog No. 02-557-805 Supplier Thermo Scientific™ Supplier No. A4101 50LA
Add to Cart
Add to Cart
The brand will transition from 3M to Thermo Scientific
Low-extractable positively charged depth filter sheets support clarification while reducing the potential for 1,3 beta-glucan interference in endotoxin testing.
  • LA Series depth filtration media is designed for low organic and inorganic extractable levels.
  • Pre-extracted cellulose fibers and filter aid help control extractables, including metallic extractables.
  • Reduced potential for 1,3 β-glucan interference in LAL endotoxin testing.
  • Positive electrokinetic charge supports adsorption of fine and submicron contaminants.
  • Combines adsorption with mechanical depth filtration for process clarification.
  • Media disc format supports evaluation and small-scale process studies.
Specifications
Quantity 40/Cs.
Type Media Sheet
Length (English) 40.15 in.