Learn More
3M Zeta Plus™ LP Series Filter Media Disc
Description
- High contaminant holding capacity for economical filtration and reliable particle removal
- Has FDA Drug Master File and USP Class VI Biological safety which eases validation and regulatory submissions by provision of documentation and traceability
- Medium is non-pyrogenic and is less likely to cause false positive tests
- LP media adsorbs contaminants too small for removal by mechanical sieving
- Self-contained, single-use disposable capsule and cartridge modules for reduced labor time for changeouts and elimination of cleaning validation.
- Tested and optimized for pharmaceutical and biological service.
Specifications
Specifications
| Disposable | Single-use |
| Format | Disc |
| Material | Cellulose |
| Sterility | Non-sterile |
| Grade | 30LP |
| Quantity | 100/Cs. |
| Type | Filtration Disc |
| Length (English) | 11.5 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.