Learn More
SOLVENTUM 3M™ Zeta Plus™ Encapsulated System Scale-Up Filter Capsule with ZB Series Media
Supplier:  SOLVENTUM 7100024302
Encompass_Preferred
Suitable for primary cell culture filtration or intermediate polishing applications in R&D, Process Development or manufacturing of recombinant protein drugs such as monoclonal antibodies. Made using cellulose fibers, diatomaceous earth and a binding resin that imparts a positive charge (quaternary amine), ZB grade media filters particles from process solutions through mechanical sieving and electrokinetic adsorption mechanisms.Wide range of filter pore sizes (0.1 to 4 µm) and a dual layer lenticular construction allows for optimizing filter throughput and filtrate quality.Scaleable single use filter portfolio enables using the same filtration platform process liquids volumes of 0.5 to 20,000 liters.
Catalog No. NC1260367