Learn More
Malvern Panalytical Zeta Potential Transfer Standard, 10 single-use syringes, 40 mV ± 5.8 mV
Catalog No. NC3724982
Encompass
Zeta Potential Transfer Standard, 10 single-use syringes, 40 mV ± 5.8 mV
Catalog No. NC3724982 Supplier Malvern Panalytical Supplier No. ZTS1240
Zeta Potential Transfer Standard, 10 single-use syringes, 40 mV ± 5.8 mV