Learn More
Medchemexpress LLC Zeteletinib | 2216753-97-6 | 98.3% | 500.47 g·mol⁻¹ | C25H23F3N4O4 | 5 MG
SDP
Catalog No. 5000220965
Encompass_Preferred
Zeteletinib is an orally active, selective RET kinase inhibitor used as a research compound for biochemical and cellular studies of RET-driven signaling. It is provided as a solid with a reported high purity and recommended cold-storage conditions to preserve stability.
  • Selective RET kinase inhibitor with nanomolar potency.
  • High selectivity versus VEGFR2 and related kinases.
  • Supplied as a solid with reported purity of 98.31%.
  • Available in small research quantities, including 5 mg and DMSO solutions.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-20965 Supplier Medchemexpress LLC Supplier No. HY1395905MG
Zeteletinib is an orally active, selective RET kinase inhibitor used as a research compound for biochemical and cellular studies of RET-driven signaling. It is provided as a solid with a reported high purity and recommended cold-storage conditions to preserve stability.
  • Selective RET kinase inhibitor with nanomolar potency.
  • High selectivity versus VEGFR2 and related kinases.
  • Supplied as a solid with reported purity of 98.31%.
  • Available in small research quantities, including 5 mg and DMSO solutions.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (6 months) or -20°C (1 month).