Learn More
Medchemexpress LLC Zeteletinib (BOS-172738; DS-5010) | 2216753-97-6 | 98.3% | 500.47 | 50 MG
SDP
Catalog No. 5000348931
Encompass_Preferred
Zeteletinib is an orally active, selective RET kinase inhibitor. It exhibits nanomolar potency against RET and demonstrates over 300-fold selectivity against VEGFR2, showing potent antitumor activity.
  • Highly potent against wild-type RET.
  • Effective against RETV804M/L gatekeeper mutants.
  • Targets the common oncogenic RET mutation M918T.
Catalog No. 50-003-48931 Supplier Medchemexpress LLC Supplier No. HY13959050MG
Zeteletinib is an orally active, selective RET kinase inhibitor. It exhibits nanomolar potency against RET and demonstrates over 300-fold selectivity against VEGFR2, showing potent antitumor activity.
  • Highly potent against wild-type RET.
  • Effective against RETV804M/L gatekeeper mutants.
  • Targets the common oncogenic RET mutation M918T.