Learn More
Cole-Parmer ZG1 High-Temp Gloves, 1000 F, 14"L, One-Size; One Pair
Supplier:  Cole-Parmer 8636400
Encompass_Preferred
The high-temperature gloves and finger cots are made of a durable cotton-backed, 11-ounce wool liner. Liner is sewn at the fingertips and double stitched with aramid thread throughout for strength and durability. One size fits all. Made in the USA.Compliance:European EN407 industrial standard
Catalog No. NC0020165
Add to Cart
This item is not returnable. View return policy
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov