Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zetomipzomib maleate 100mg | 2170983-62-5 | 100 MG
SDP

Catalog No. 5000229071
Encompass_Preferred

Zetomipzomib maleate is the maleate salt of zetomipzomib, a proteasome inhibitor supplied for laboratory research use. It is provided as a solid reagent for preclinical and biochemical studies, and is intended for research purposes only.

  • Maleate salt of a proteasome inhibitor.
  • High purity (99.8%) suitable for analytical and biological assays.
  • Molecular formula C34H46N4O12; molecular weight 702.75.
  • Packaged as a 100 mg solid for laboratory handling and storage.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-29071 Supplier Medchemexpress LLC Supplier No. HY114419A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Zetomipzomib maleate is the maleate salt of zetomipzomib, a proteasome inhibitor supplied for laboratory research use. It is provided as a solid reagent for preclinical and biochemical studies, and is intended for research purposes only.

  • Maleate salt of a proteasome inhibitor.
  • High purity (99.8%) suitable for analytical and biological assays.
  • Molecular formula C34H46N4O12; molecular weight 702.75.
  • Packaged as a 100 mg solid for laboratory handling and storage.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.