Learn More
Medchemexpress LLC Zetomipzomib maleate 1mg | 2170983-62-5 | 1 MG
SDP
Catalog No. 5000229728
Encompass_Preferred
Zetomipzomib maleate is a selective immunoproteasome inhibitor (KZR-616 maleate) used as a research reagent to study autoimmune and inflammatory pathways. It is supplied as a small, research-scale sample suitable for in vitro and preclinical work.
  • Selective inhibitor of immunoproteasome subunits LMP7 and LMP2.
  • Suitable for preclinical research and mechanistic studies.
  • Provided as a low-amount research sample for laboratory use.
  • Stored and handled according to reagent safety protocols and SDS guidance.
Catalog No. 50-002-29728 Supplier Medchemexpress LLC Supplier No. HY114419A1MG
Zetomipzomib maleate is a selective immunoproteasome inhibitor (KZR-616 maleate) used as a research reagent to study autoimmune and inflammatory pathways. It is supplied as a small, research-scale sample suitable for in vitro and preclinical work.
  • Selective inhibitor of immunoproteasome subunits LMP7 and LMP2.
  • Suitable for preclinical research and mechanistic studies.
  • Provided as a low-amount research sample for laboratory use.
  • Stored and handled according to reagent safety protocols and SDS guidance.