Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zetomipzomib maleate 25mg | 2170983-62-5 | 25 MG
SDP

Catalog No. 5000308049
Encompass_Preferred

Zetomipzomib maleate is an immunoproteasome-selective inhibitor used in preclinical research to study proteasome biology and mechanisms of autoimmune disease. It is provided as a high-purity solid suitable for in vitro assays and biochemical studies.

  • Immunoproteasome-selective inhibitor targeting LMP7 and LMP2.
  • High purity suitable for research (99.8%).
  • Available in small-scale pack sizes for laboratory use.
  • Molecular formula C34H46N4O12, molecular weight 702.75.

Catalog No. 50-003-08049 Supplier Medchemexpress LLC Supplier No. HY114419A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Zetomipzomib maleate is an immunoproteasome-selective inhibitor used in preclinical research to study proteasome biology and mechanisms of autoimmune disease. It is provided as a high-purity solid suitable for in vitro assays and biochemical studies.

  • Immunoproteasome-selective inhibitor targeting LMP7 and LMP2.
  • High purity suitable for research (99.8%).
  • Available in small-scale pack sizes for laboratory use.
  • Molecular formula C34H46N4O12, molecular weight 702.75.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.