Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zetomipzomib maleate 50mg | 2170983-62-5 | 50 MG
SDP

Catalog No. 5000308130
Encompass_Preferred

Zetomipzomib maleate is an investigational immunoproteasome inhibitor supplied for laboratory research use. It selectively targets immunoproteasome subunits and is used in studies of proteasome biology and autoimmune disease models.

  • Selective immunoproteasome inhibition (LMP7, LMP2) for targeted proteasome research.
  • Reported molecular weight 702.75 g/mol and formula C34H46N4O12.
  • Soluble in DMSO and water; multiple in vivo formulation options provided.
  • Recommended sealed storage and specified solvent storage conditions.
  • For research use only; not for clinical or human therapeutic use.

Catalog No. 50-003-08130 Supplier Medchemexpress LLC Supplier No. HY114419A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Zetomipzomib maleate is an investigational immunoproteasome inhibitor supplied for laboratory research use. It selectively targets immunoproteasome subunits and is used in studies of proteasome biology and autoimmune disease models.

  • Selective immunoproteasome inhibition (LMP7, LMP2) for targeted proteasome research.
  • Reported molecular weight 702.75 g/mol and formula C34H46N4O12.
  • Soluble in DMSO and water; multiple in vivo formulation options provided.
  • Recommended sealed storage and specified solvent storage conditions.
  • For research use only; not for clinical or human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.