Learn More
MSC AlorAir Air Mover, 950 CFM, 115 VAC, 8-Speed Control, ETL Certified
Catalog No. 502840063
Encompass
This air mover is a commercial blower designed for various uses including carpets, walls, and plumbing. It features variable speed control and delivers 950 CFM airflow with 1.8 Amps of power.
  • Circuit breaker protector and GFCI outlet for safety
  • 8-speed control and 5 drying positions for versatility
  • Over 900 CFM airflow with less than 1.8 amps power consumption
  • Lightweight, portable design with flex handle and 20 ft AC cord
  • Units interlock for stacking; daisy chain up to 6 fans
  • Rugged steel body and chemically resistant, washable housing
  • Automatic hour meter and LCD display
  • Aids in salvaging water-damaged possessions and improving indoor air quality
  • Optimizes restoration jobs and protects properties from moisture damage
  • Provides clean, safe ventilation for increased productivity
Catalog No. 50-284-0063 Supplier MSC Supplier No. 86603164
This air mover is a commercial blower designed for various uses including carpets, walls, and plumbing. It features variable speed control and delivers 950 CFM airflow with 1.8 Amps of power.
  • Circuit breaker protector and GFCI outlet for safety
  • 8-speed control and 5 drying positions for versatility
  • Over 900 CFM airflow with less than 1.8 amps power consumption
  • Lightweight, portable design with flex handle and 20 ft AC cord
  • Units interlock for stacking; daisy chain up to 6 fans
  • Rugged steel body and chemically resistant, washable housing
  • Automatic hour meter and LCD display
  • Aids in salvaging water-damaged possessions and improving indoor air quality
  • Optimizes restoration jobs and protects properties from moisture damage
  • Provides clean, safe ventilation for increased productivity