Learn More
ATCC ZF4; Embryo; Zebrafish (Danio rerio)
Supplier:  ATCC CRL2050
Encompass
ZF4 is a fibroblast cell line that was isolated from the embryo of a zebrafish. This cell line was deposited by W Driever.
Catalog No. 50-238-1885