Learn More
Aviva Systems Biology Corp ZFHX1B Antibody - N-terminal region (ARP33694_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33694_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZEB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZFHX1B. Alternate names: KIAA0569, SIP-1, SIP1, SMADIP1, ZFHX1B, ZEB2, HSPC082ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-52918