Learn More
Aviva Systems Biology Corp ZFP1 Antibody - N-terminal region (ARP31586_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31586_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFP1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ZFP1. Alternate names: ZNF475ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Rat
Catalog No. 50-014-52831