Learn More
Aviva Systems Biology Corp Zfp109 Antibody - C-terminal region (ARP38428_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38428_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zfp109. It was validated on Western Blot. . Alternate names: MGC141452ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-733