Learn More
Aviva Systems Biology Corp Zfp113 Antibody - middle region (ARP50471_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50471_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zfp113. It was validated on Western Blot. . Alternate names: 4732456B05Rik, KIAA4229, mKIAA4229ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-510