Learn More
Aviva Systems Biology Corp Zfp275 Antibody - middle region (ARP47780_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47780_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zfp275. It was validated on Western Blot. . Alternate names: 5430431L06Rik, AI593314, DXBay20, DXHXS52, DXPas8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-327