Learn More
Aviva Systems Biology Corp ZFP28 Antibody - middle region (ARP39445_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39445_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFP28. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZFP28. Alternate names: DKFZp686K03205, KIAA1431, MGC104540, mkr5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57032