Learn More
Aviva Systems Biology Corp Zfp354a Antibody - C-terminal region (ARP50736_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50736_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zfp354a. It was validated on Western Blot. . Alternate names: AW488485, MGC91222, Tcf17, Znf354a, kid1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-536