Learn More
Aviva Systems Biology Corp ZFP36 Antibody - N-terminal region (ARP38303_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38303_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFP36. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZFP36. Alternate names: TTP, G0S24, GOS24, TIS11, NUP475, zfp-36, RNF162AApplicationWB. Species reactivity: Human
Catalog No. 50-014-56602