Learn More
Aviva Systems Biology Corp ZFP36 Antibody - middle region (ARP38304_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38304_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFP36. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZFP36. Alternate names: G0S24, GOS24, NUP475, RNF162A, TIS11, TTPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-56603