Learn More
Aviva Systems Biology Corp ZFP36L1 Antibody - N-terminal region (ARP33381_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33381_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFP36L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZFP36L1. Alternate names: BRF1, ERF1, cMG1, ERF-1, Berg36, TIS11B, RNF162BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-54990