Learn More
Aviva Systems Biology Corp Zfp36l1 Antibody - C-terminal region (ARP57901_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57901_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zfp36l1. It was validated on Western Blot. . Alternate names: AW742437, AW743212, Berg36, Brf1, D530020L18Rik, ERF1, TIS11b, cMG1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-411