Learn More
Aviva Systems Biology Corp ZFP36L1 Antibody - middle region (ARP38524_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38524_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFP36L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZFP36L1. Alternate names: BRF1, Berg36, ERF-1, ERF1, RNF162B, TIS11B, cMG1ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56686