Learn More
Aviva Systems Biology Corp ZFP37 Antibody - middle region (ARP33495_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33495_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFP37. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZFP37. Alternate names: FLJ38524, ZNF906ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-55044