Learn More
Aviva Systems Biology Corp ZFP42 Antibody - middle region (ARP36245_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36245_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFP42. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZFP42. Alternate names: REX1, ZNF754ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56088