Learn More
Aviva Systems Biology Corp Zfp444 Antibody - C-terminal region (ARP37390_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37390_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zfp444. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2810031J10Rik, 6230401O10Rik, FLJ00134, mFLJ00134ApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-284-623