Learn More
Aviva Systems Biology Corp Zfp445 Antibody - middle region (ARP37532_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37532_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zfp445. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AW610627, ZNF168, mszf29, mszf50, Znf445ApplicationWB. Species reactivity: Mouse
Catalog No. 50-284-652