Learn More
Aviva Systems Biology Corp ZFP57 Antibody - N-terminal region (ARP40121_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40121_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFP57. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ZFP57. Alternate names: ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-57299