Learn More
Aviva Systems Biology Corp ZFP64 Antibody - middle region (ARP33156_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33156_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFP64. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZFP64. Alternate names: MGC940, ZNF338ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-54898