Learn More
Aviva Systems Biology Corp Zfp759 Antibody - N-terminal region (ARP39917_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39917_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zfp759. It was validated on Western Blot. . Alternate names: Rslcan8, BC028265, Rslcan-8ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-841