Learn More
Aviva Systems Biology Corp Zfp874b Antibody - middle region (ARP51663_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51663_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zfp874b. It was validated on Western Blot. . Alternate names: Rslcan16, 9630025I21RikApplicationWB. Species reactivity: Cow, Human, Mouse, Rat
Catalog No. 50-285-583