Learn More
Aviva Systems Biology Corp ZFP91 Antibody - middle region (ARP36120_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36120_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFP91. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZFP91. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast
Catalog No. 50-014-56023