Learn More
Aviva Systems Biology Corp ZFP91 Antibody - N-terminal region (ARP36119_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36119_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFP91. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZFP91. Alternate names: PZF, DMS-8, DSM-8, FKSG11, ZFP-91, ZNF757ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56021