Learn More
Aviva Systems Biology Corp ZFPL1 Antibody - middle region (ARP34354_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34354_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFPL1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ZFPL1. Alternate names: D11S750, MCG4ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55351