Learn More
Aviva Systems Biology Corp ZFPM1 Antibody - N-terminal region (ARP37001_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37001_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFPM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse ZFPM1. Alternate names: FOG, Fog1ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-014-56309