Learn More
Aviva Systems Biology Corp ZFR Antibody - middle region (ARP34527_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34527_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZFR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZFR. Alternate names: FLJ41312, ZFR1ApplicationWB, IHC. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55435